siRNA Details
| Gene Symbol: Jak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146145.2 | NP_666257.2 | 4 | 592 | ||
| GenBank mRNA | AK081552.1 | - | 4 | 567 | ||
| AK133866.1 | - | 4 | 532 | |||
| AK134143.1 | - | 4 | 535 | |||
| AK141210.1 | - | 4 | 534 | |||
| AK143650.1 | - | 4 | 537 | |||
| AK148521.1 | - | 4 | 557 | |||
| AK152128.1 | - | 4 | 555 | |||
| BC039752.1 | - | 4 | 561 | |||
| S63728.1 | - | 4 | 542 | |||

Jak1
Genomic Map