siRNA Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048177.2 | NP_001041642.1 | 22 | 3416 | ||
| NM_008413.3 | NP_032439.2 | 23 | 3499 | |||
| GenBank mRNA | BC054807.1 | - | 22 | 3179 | 6838318 | |
| BC059834.1 | - | 22 | 3179 | 6838318 | ||
| M33423.1 | - | 2 | 161 | |||

Jak2
Genomic Map