siRNA Details
| Gene Symbol: Atcay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178662.3 | NP_848777.1 | 4 | 510 | ||
| GenBank mRNA | AK028298.1 | - | 4 | 517 | ||
| AK038483.1 | - | 4 | 520 | |||
| AK038990.1 | - | 4 | 511 | |||
| AK039411.1 | - | 4 | 518 | |||
| AK086723.1 | - | 4 | 552 | |||
| AK162959.1 | - | 4 | 518 | |||
| AY349150.1 | - | 4 | 475 | |||
| BC048903.1 | - | 4 | 494 | 6491141 | ||

Atcay
Genomic Map