siRNA Details
| Gene Symbol: Jarid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205043.1 | NP_001191972.1 | 4 | 953 | ||
| NM_001205044.1 | NP_001191973.1 | 5 | 626 | |||
| NM_021878.3 | NP_068678.1 | 5 | 672 | |||
| GenBank mRNA | AK044888.1 | - | 4 | 857 | ||
| AK045214.1 | - | 4 | 758 | |||
| AK147226.1 | - | 4 | 953 | |||
| AK164134.1 | - | 3 | 277 | |||
| BC026056.1 | - | 4 | 860 | |||
| BC050129.1 | - | 4 | 874 | |||
| BC051010.1 | - | 4 | 856 | |||
| BC052444.1 | - | 5 | 543 | 6406875 | ||
| BC060695.1 | - | 5 | 543 | 6406875 | ||
| D31967.1 | - | 5 | 562 | |||

Jarid2
Genomic Map