siRNA Details
| Gene Symbol: Ajuba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010590.5 | NP_034720.2 | 5, 6 | 1790 | ||
| GenBank mRNA | AK012077.1 | - | 3, 4 | 507 | ||
| BC011035.1 | - | 5, 6 | 1551 | 4187633 | ||
| U79776.1 | - | 5, 6 | 1659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ajuba| Gene Symbol: Ajuba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010590.5 | NP_034720.2 | 5, 6 | 1790 | ||
| GenBank mRNA | AK012077.1 | - | 3, 4 | 507 | ||
| BC011035.1 | - | 5, 6 | 1551 | 4187633 | ||
| U79776.1 | - | 5, 6 | 1659 | |||