siRNA Details
| Gene Symbol: HOXD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014213.3 | NP_055028.3 | 2 | 1091 | ||
| GenBank mRNA | BC044855.1 | - | 2 | 1091 | 5191006 | |
| X59372.1 | - | 2 | 1013 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD9| Gene Symbol: HOXD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014213.3 | NP_055028.3 | 2 | 1091 | ||
| GenBank mRNA | BC044855.1 | - | 2 | 1091 | 5191006 | |
| X59372.1 | - | 2 | 1013 | |||