siRNA Details
| Gene Symbol: HOXD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002148.3 | NP_002139.2 | 2 | 891 | ||
| GenBank mRNA | BC069619.1 | - | 2 | 871 | 7262455 | |
| BC074760.2 | - | 2 | 870 | 30915219 | ||
| BX114645.1 | - | 2 | 259 | |||
| X59373.1 | - | 2 | 854 | |||

HOXD10
Genomic Map