siRNA Details
Gene Symbol: Kcna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010596.2 | NP_034726.2 | 2 | 1437 | ||
GenBank mRNA | AF032099.1 | - | 2 | 1164 | ||
AY779767.1 | - | 2 | 1171 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010596.2 | NP_034726.2 | 2 | 1437 | ||
GenBank mRNA | AF032099.1 | - | 2 | 1164 | ||
AY779767.1 | - | 2 | 1171 |