siRNA Details
Gene Symbol: HOXD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021192.2 | NP_067015.2 | 2 | 871 | ||
GenBank mRNA | AK309681.1 | - | 2 | 195 | ||
BC109394.1 | - | 3 | 373 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOXD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021192.2 | NP_067015.2 | 2 | 871 | ||
GenBank mRNA | AK309681.1 | - | 2 | 195 | ||
BC109394.1 | - | 3 | 373 |