siRNA Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252654.1 | NP_001239583.1 | 14 | 1164 | ||
| NM_001252655.1 | NP_001239584.1 | 14 | 1193 | |||
| NM_001252656.1 | NP_001239585.1 | 15 | 1235 | |||
| NM_010598.3 | NP_034728.2 | 15 | 1206 | |||
| GenBank mRNA | AK143166.1 | - | 14 | 1141 | ||
| AK157053.1 | - | 14 | 1157 | |||
| BC039178.1 | - | 15 | 1136 | 4503373 | ||
| L48983.1 | - | 15 | 1198 | |||
| U31908.1 | - | 15 | 1143 | |||
| U65592.1 | - | 14 | 1001 | |||

Kcnab2
Genomic Map