siRNA Details
| Gene Symbol: Kcnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112739.2 | NP_001106210.1 | 2 | 2043 | ||
| NM_008421.3 | NP_032447.1 | 2 | 2043 | |||
| GenBank mRNA | AK147476.1 | - | 2 | 2016 | ||
| AK147682.1 | - | 2 | 2016 | |||
| AK162964.1 | - | 2 | 700 | |||
| BC125470.1 | - | 2 | 880 | 40129985 | ||
| BC132439.1 | - | 2 | 880 | 40130716 | ||
| Y07521.1 | - | 2 | 969 | |||

Kcnc1
Genomic Map