siRNA Details
| Gene Symbol: Kcnh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010601.3 | NP_034731.3 | 3 | 337 | ||
| GenBank mRNA | AY380579.1 | - | 3 | 337 | ||
| BC141013.1 | - | 3 | 452 | 9055625 | ||
| BC145145.1 | - | 3 | 452 | 9053696 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh3| Gene Symbol: Kcnh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010601.3 | NP_034731.3 | 3 | 337 | ||
| GenBank mRNA | AY380579.1 | - | 3 | 337 | ||
| BC141013.1 | - | 3 | 452 | 9055625 | ||
| BC145145.1 | - | 3 | 452 | 9053696 | ||