siRNA Details
| Gene Symbol: Kcnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010608.2 | NP_034738.1 | 2 | 752 | ||
| GenBank mRNA | AB008537.1 | - | 2 | 618 | ||
| AB013345.1 | - | 2 | 740 | |||
| AF006824.1 | - | 2 | 596 | |||
| AF022821.1 | - | 2 | 599 | |||
| AF065162.1 | - | 2 | 836 | |||
| AI325858.1 | - | 2 | 410 | |||
| BC032047.1 | - | 2 | 752 | |||

Kcnk3
Genomic Map