siRNA Details
| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008431.2 | NP_032457.1 | 2 | 377 | ||
| GenBank mRNA | AF056492.1 | - | 2 | 415 | ||
| BC119784.1 | - | 2 | 247 | 40106379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk4| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008431.2 | NP_032457.1 | 2 | 377 | ||
| GenBank mRNA | AF056492.1 | - | 2 | 415 | ||
| BC119784.1 | - | 2 | 247 | 40106379 | ||