siRNA Details
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008431.2 | NP_032457.1 | 3 | 479 | ||
GenBank mRNA | AF056492.1 | - | 3 | 517 | ||
BC119784.1 | - | 3 | 349 | 40106379 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008431.2 | NP_032457.1 | 3 | 479 | ||
GenBank mRNA | AF056492.1 | - | 3 | 517 | ||
BC119784.1 | - | 3 | 349 | 40106379 |