siRNA Details
| Gene Symbol: Kcnmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031169.4 | NP_112446.2 | 3 | 494 | ||
| GenBank mRNA | AF020711.1 | - | 3 | 494 | ||
| AJ001291.1 | - | 3 | 236 | |||
| AK038987.1 | - | 3 | 222 | |||
| BC013338.1 | - | 3 | 285 | 3988005 | ||

Kcnmb1
Genomic Map