siRNA Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163510.1 | NP_001156982.1 | 7 | 1236 | ||
| NM_008433.4 | NP_032459.3 | 7 | 1236 | |||
| GenBank mRNA | AB128930.1 | - | 5 | 999 | ||
| AF042487.1 | - | 7 | 1102 | |||
| AF072884.1 | - | 7 | 1062 | |||
| AK010943.1 | - | 7 | 1237 | |||
| AK085752.1 | - | 7 | 1053 | |||
| AK088778.1 | - | 2 | 160 | |||
| AK155910.1 | - | 7 | 1201 | |||
| BC010274.1 | - | 7 | 1185 | 3589411 | ||

Kcnn4
Genomic Map