siRNA Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008434.2 | NP_032460.2 | 9 | 1285 | ||
| GenBank mRNA | AB079603.1 | - | 9 | 988 | ||
| AK136389.1 | - | 2 | 155 | |||
| AK154244.1 | - | 9 | 1259 | |||
| AY331142.1 | - | 9 | 1200 | |||
| AY644390.1 | - | 4 | 393 | |||
| BC045142.1 | - | 9 | 1285 | 6314642 | ||
| BC055304.1 | - | 9 | 1240 | 6397978 | ||
| U70068.1 | - | 9 | 1091 | |||

Kcnq1
Genomic Map