siRNA Details
| Gene Symbol: Kdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010612.2 | NP_034742.2 | 3, 4 | 629 | ||
| GenBank mRNA | AK054510.1 | - | 3, 4 | 662 | ||
| AK141938.1 | - | 3, 4 | 652 | |||
| BC020530.1 | - | 3, 4 | 629 | 4238984 | ||
| EU884114.1 | - | 3, 4 | 659 | |||
| S53103.1 | - | 3, 4 | 640 | |||
| X59397.1 | - | 3, 4 | 576 | |||
| X70842.1 | - | 3, 4 | 654 | |||

Kdr
Genomic Map