siRNA Details
| Gene Symbol: Kdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010612.2 | NP_034742.2 | 11 | 1696 | ||
| GenBank mRNA | AK054510.1 | - | 10 | 2592 | ||
| AK141938.1 | - | 11 | 1719 | |||
| BC020530.1 | - | 11 | 1696 | 4238984 | ||
| EU884114.1 | - | 11 | 1726 | |||
| S53103.1 | - | 11 | 1707 | |||
| X59397.1 | - | 11 | 1643 | |||
| X70842.1 | - | 11 | 1721 | |||

Kdr
Genomic Map