siRNA Details
| Gene Symbol: Kif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110315.2 | NP_001103785.1 | 30 | 3319 | ||
| NM_001294149.1 | NP_001281078.1 | 30 | 3319 | |||
| NM_001294150.1 | NP_001281079.1 | 30 | 3234 | |||
| NM_008440.4 | NP_032466.2 | 30 | 3319 | |||
| GenBank mRNA | AK147640.1 | - | 30 | 3277 | ||
| AK220487.1 | - | 15 | 1495 | |||
| AY426748.1 | - | 30 | 3175 | |||
| BC054538.1 | - | 30 | 3291 | |||
| BC062891.1 | - | 30 | 3298 | 5702372 | ||
| D29951.1 | - | 30 | 3268 | |||

Kif1a
Genomic Map