siRNA Details
Gene Symbol: Kif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290995.1 | NP_001277924.1 | 15 | 1716 | ||
NM_008441.2 | NP_032467.2 | 13 | 1543 | |||
NM_207682.2 | NP_997565.2 | 13 | 1543 | |||
GenBank mRNA | AB023656.1 | - | 13 | 1427 | ||
AF090190.1 | - | 14 | 1342 | |||
AF131865.1 | - | 15 | 1634 | |||
AK134946.1 | - | 13 | 1579 | |||
AK147409.1 | - | 3 | 230 | |||
D17577.1 | - | 13 | 1496 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
