siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 22 | 2676 | ||
| GenBank mRNA | AB074017.1 | - | 20 | 2306 | ||
| AK046574.1 | - | 17 | 1917 | |||
| AK173007.1 | - | 21 | 2419 | |||
| BC006188.1 | - | 4 | 624 | |||
| BC029286.1 | - | 22 | 2663 | |||

Kif1c
Genomic Map