siRNA Details
| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145779.1 | NP_001139251.1 | 19 | 2007 | ||
| NM_008442.2 | NP_032468.2 | 21 | 2504 | |||
| GenBank mRNA | AK037037.1 | - | 19 | 1920 | ||
| BB622253.1 | - | 3 | 277 | |||
| BC006803.1 | - | 19 | 2002 | 3152883 | ||
| D12644.1 | - | 21 | 2505 | |||
| Y15894.1 | - | 18 | 1857 | |||

Kif2a
Genomic Map