siRNA Details
| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145779.1 | NP_001139251.1 | 11 | 1037 | ||
| NM_008442.2 | NP_032468.2 | 12 | 1420 | |||
| GenBank mRNA | AK037037.1 | - | 11 | 950 | ||
| BC006803.1 | - | 11 | 1032 | 3152883 | ||
| D12644.1 | - | 12 | 1421 | |||
| Y15894.1 | - | 10 | 887 | |||

Kif2a
Genomic Map