siRNA Details
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 1 | 1068 | ||
GenBank mRNA | AF013116.1 | - | 1 | 644 | ||
AK133649.1 | - | 1 | 1050 | |||
AK147572.1 | - | 1 | 870 | |||
AK220283.1 | - | 2 | 793 | |||
BC052732.1 | - | 1 | 225 | |||
BC053053.1 | - | 1 | 1010 | |||
BC062648.1 | - | 1 | 743 | |||
BC127063.1 | - | 1 | 280 | 40109965 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
