siRNA Details
| Gene Symbol: Kif4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008446.2 | NP_032472.2 | 28, 29 | 3784 | ||
| GenBank mRNA | AK084837.1 | - | 15, 16 | 2120 | ||
| AK135430.1 | - | 28, 29 | 3279 | |||
| AK136346.1 | - | 9, 10 | 1025 | |||
| AK171938.1 | - | 28, 29 | 3696 | |||
| BC050946.1 | - | 28, 29 | 3722 | 6822283 | ||
| BC054537.1 | - | 28, 29 | 3706 | |||
| D12646.1 | - | 28, 29 | 4111 | |||

Kif4
Genomic Map