siRNA Details
| Gene Symbol: Kif5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008449.2 | NP_032475.2 | 4, 5 | 737 | ||
| GenBank mRNA | AB093244.1 | - | 4, 5 | 630 | ||
| AF067180.1 | - | 4, 5 | 389 | |||
| AK158884.1 | - | 4, 5 | 723 | |||
| BC067051.1 | - | 4, 5 | 737 | 30536079 | ||

Kif5c
Genomic Map