siRNA Details
| Gene Symbol: Kifc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145831.1 | NP_001139303.1 | 5, 6 | 638 | ||
| NM_010631.2 | NP_034761.3 | 3, 4 | 465 | |||
| GenBank mRNA | AK166482.1 | - | 5, 6 | 616 | ||
| AK186859.1 | - | 1, 2 | 41 | |||
| AK189996.1 | - | 2, 3 | 174 | |||
| BC070429.1 | - | 3, 4 | 465 | 6810893 | ||
| BI737872.1 | - | 1, 2 | 57 | |||
| D49546.1 | - | 3, 4 | 480 | |||

Kifc3
Genomic Map