siRNA Details
| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025280.2 | NP_079556.1 | 5 | 1125 | ||
| GenBank mRNA | AK132548.1 | - | 5 | 1669 | ||
| AK167740.1 | - | 5 | 1129 | |||
| BC028860.1 | - | 5 | 1125 | 3673830 | ||
| BC058169.1 | - | 4 | 2508 | |||
| X58472.1 | - | 5 | 1130 | |||

Kin
Genomic Map