siRNA Details
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272097.1 | NP_001259026.1 | 2 | 276 | ||
NM_010634.3 | NP_034764.1 | 2 | 276 | |||
GenBank mRNA | AK008782.1 | - | 2 | 270 | ||
AK011551.1 | - | 2 | 272 | |||
AK166506.1 | - | 2 | 271 | |||
AK167389.1 | - | 2 | 271 | |||
AK216545.1 | - | 1 | 85 | |||
BC002008.1 | - | 2 | 256 | 3490535 | ||
BC053042.1 | - | 1 | 2508 | |||
BC100543.1 | - | 2 | 251 | 30136900 | ||
X70100.1 | - | 2 | 255 |
