siRNA Details
| Gene Symbol: Klf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010636.3 | NP_034766.2 | 7 | 1115 | ||
| GenBank mRNA | AB221575.1 | - | 7 | 1083 | ||
| AK042450.1 | - | 2 | 367 | |||
| BC067408.1 | - | 7 | 1115 | 30091275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf12| Gene Symbol: Klf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010636.3 | NP_034766.2 | 7 | 1115 | ||
| GenBank mRNA | AB221575.1 | - | 7 | 1083 | ||
| AK042450.1 | - | 2 | 367 | |||
| BC067408.1 | - | 7 | 1115 | 30091275 | ||