siRNA Details
| Gene Symbol: Klf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008452.2 | NP_032478.2 | 3 | 1166 | ||
| GenBank mRNA | AK131962.1 | - | 3 | 1167 | ||
| BC138737.1 | - | 3 | 1174 | 8861759 | ||
| BC138738.1 | - | 3 | 1174 | 8861760 | ||
| U25096.1 | - | 3 | 1195 | |||

Klf2
Genomic Map