siRNA Details
| Gene Symbol: HPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020995.3 | NP_066275.3 | 3 | 194 | ||
| GenBank mRNA | AV698541.1 | - | 3 | 179 | ||
| BM547990.1 | - | 1 | 81 | |||
| CB115435.1 | - | 3 | 193 | |||
| CB147217.1 | - | 3 | 194 | |||
| T71114.1 | - | 3 | 179 | |||

HPR
Genomic Map