siRNA Details
| Gene Symbol: Serpina3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008458.2 | NP_032484.1 | 3 | 905 | ||
| GenBank mRNA | BC116724.1 | - | 3 | 903 | 40106394 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina3c| Gene Symbol: Serpina3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008458.2 | NP_032484.1 | 3 | 905 | ||
| GenBank mRNA | BC116724.1 | - | 3 | 903 | 40106394 | |