siRNA Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016659.3 | NP_057868.2 | 9 | 1608 | ||
| GenBank mRNA | AF074456.1 | - | 7 | 1223 | ||
| AK080158.1 | - | 7 | 1190 | |||
| AK172530.1 | - | 1091 | ||||
| M25775.1 | - | 7 | 1212 | |||
| M25812.1 | - | 7 | 1146 | |||

Klra1
Genomic Map