siRNA Details
Gene Symbol: Klra13-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074460.1 | - | 3 | 381 | ||
AF283252.1 | - | 382 | ||||
BC099861.1 | - | 296 | 40040023 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klra13-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074460.1 | - | 3 | 381 | ||
AF283252.1 | - | 382 | ||||
BC099861.1 | - | 296 | 40040023 |