siRNA Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170851.1 | NP_001164322.1 | 8 | 956 | ||
| NM_008462.5 | NP_032488.4 | 7 | 857 | |||
| GenBank mRNA | AF253057.1 | - | 6 | 691 | ||
| AF253058.1 | - | 6 | 691 | |||
| AF253059.1 | - | 6 | 691 | |||
| AF395446.1 | - | 6 | 691 | |||
| AK017140.1 | - | 7 | 848 | |||
| AY860971.1 | - | 6 | 691 | |||
| BC064711.1 | - | 8 | 857 | 30295755 | ||
| EF025059.1 | - | 8 | 790 | |||
| U10304.1 | - | 7 | 785 | |||

Klra2
Genomic Map