siRNA Details
| Gene Symbol: Klra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008463.2 | NP_032489.1 | 4 | 344 | ||
| GenBank mRNA | AF247643.1 | - | 4 | 313 | ||
| AF419251.1 | - | 477 | ||||
| AY620247.1 | - | 3 | 236 | |||
| BC120550.1 | - | 4 | 302 | 40129473 | ||
| BC131790.1 | - | 4 | 286 | 40101177 | ||
| BC137708.1 | - | 4 | 302 | 8860727 | ||
| FJ159531.1 | - | 3 | 236 | |||
| U10091.1 | - | 4 | 344 | |||

Klra5
Genomic Map