siRNA Details
| Gene Symbol: Mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010658.3 | NP_034788.1 | 1 | 935 | ||
| GenBank mRNA | AK131926.1 | - | 1 | 81 | ||
| AK132425.1 | - | 1 | 936 | |||
| AK143209.1 | - | 1 | 937 | |||
| AK154830.1 | - | 1 | 297 | |||
| BC016434.1 | - | 1 | 638 | 4485468 | ||
| BC038256.1 | - | 1 | 883 | 5097650 | ||
| L36435.1 | - | 1 | 715 | |||

Mafb
Genomic Map