siRNA Details
| Gene Symbol: Krt34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027563.3 | NP_081839.1 | 7 | 1279 | ||
| GenBank mRNA | AK014631.1 | - | 7 | 1281 | ||
| BC125420.1 | - | 7 | 1257 | 40129935 | ||
| BC125446.1 | - | 7 | 1257 | 40129961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt34| Gene Symbol: Krt34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027563.3 | NP_081839.1 | 7 | 1279 | ||
| GenBank mRNA | AK014631.1 | - | 7 | 1281 | ||
| BC125420.1 | - | 7 | 1257 | 40129935 | ||
| BC125446.1 | - | 7 | 1257 | 40129961 | ||