siRNA Details
| Gene Symbol: Krt84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008474.2 | NP_032500.2 | 5 | 1081 | ||
| GenBank mRNA | AY028607.1 | - | 5 | 1038 | ||
| BC114971.1 | - | 5 | 1034 | 40039508 | ||
| CJ052880.1 | - | 3 | 310 | |||
| X65505.1 | - | 2 | 101 | |||

Krt84
Genomic Map