siRNA Details
| Gene Symbol: Krt6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010669.2 | NP_034799.2 | 9 | 1999 | ||
| GenBank mRNA | AK009241.1 | - | 6 | 1105 | ||
| AK137688.1 | - | 9 | 1998 | |||
| DV553143.1 | - | 1 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt6b| Gene Symbol: Krt6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010669.2 | NP_034799.2 | 9 | 1999 | ||
| GenBank mRNA | AK009241.1 | - | 6 | 1105 | ||
| AK137688.1 | - | 9 | 1998 | |||
| DV553143.1 | - | 1 | 65 | |||