siRNA Details
| Gene Symbol: Krtap6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010673.2 | NP_034803.2 | 1 | 640 | ||
| GenBank mRNA | AA763314.1 | - | 1 | 135 | ||
| BC116356.1 | - | 1 | 639 | |||
| BC116357.1 | - | 1 | 639 | |||
| BC128485.1 | - | 1 | 639 | |||
| BC128486.1 | - | 1 | 639 | |||

Krtap6-2
Genomic Map