siRNA Details
| Gene Symbol: Krtap19-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010676.2 | NP_034806.2 | 1 | 129 | ||
| GenBank mRNA | AA717139.1 | - | 1 | 137 | ||
| D86422.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap19-5| Gene Symbol: Krtap19-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010676.2 | NP_034806.2 | 1 | 129 | ||
| GenBank mRNA | AA717139.1 | - | 1 | 137 | ||
| D86422.1 | - | 1 | 140 | |||