siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 14, 15 | 2090 | ||
| GenBank mRNA | AK137532.1 | - | 13, 14 | 1827 | ||
| U43585.1 | - | 14, 15 | 2097 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ksr1| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 14, 15 | 2090 | ||
| GenBank mRNA | AK137532.1 | - | 13, 14 | 1827 | ||
| U43585.1 | - | 14, 15 | 2097 | |||