siRNA Details
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293635.1 | NP_001280564.1 | 2, 3 | 681 | ||
| NM_008477.2 | NP_032503.2 | 1, 2 | 552 | |||
| GenBank mRNA | BC046994.1 | - | 2, 3 | 682 | ||
| BC066871.1 | - | 2, 3 | 636 | |||
| CO431187.1 | - | 1, 2 | 389 | |||
| L43326.1 | - | 1, 2 | 540 | |||

Ktn1
Genomic Map