siRNA Details
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293635.1 | NP_001280564.1 | 2 | 414 | ||
| NM_001293636.1 | NP_001280565.1 | 3 | 539 | |||
| NM_008477.2 | NP_032503.2 | 1 | 285 | |||
| GenBank mRNA | AK040200.1 | - | 2 | 415 | ||
| BC046994.1 | - | 2 | 415 | |||
| BC066871.1 | - | 2 | 368 | |||
| BC075697.1 | - | 3 | 456 | |||
| CO431187.1 | - | 1 | 122 | |||
| L43326.1 | - | 1 | 273 | |||

Ktn1
Genomic Map