siRNA Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 9 | 1304 | ||
| GenBank mRNA | L20477.1 | - | 7 | 864 | ||
| U43327.2 | - | 8 | 1037 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamc2| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 9 | 1304 | ||
| GenBank mRNA | L20477.1 | - | 7 | 864 | ||
| U43327.2 | - | 8 | 1037 | |||