siRNA Details
| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008486.2 | NP_032512.2 | 17 | 2484 | ||
| GenBank mRNA | AK143746.1 | - | 17 | 2427 | ||
| AK171427.1 | - | 18 | 2846 | |||
| BC005431.1 | - | 17 | 2450 | 3593307 | ||
| BC017011.1 | - | 17 | 2508 | 3590117 | ||
| BC040792.1 | - | 17 | 2508 | 3590117 | ||
| U77083.1 | - | 17 | 2416 | |||

Anpep
Genomic Map